Phenoxybenzamine hydrochloride, Thermo Scientific Chemicals
Phenoxybenzamine hydrochloride, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Phenoxybenzamine hydrochloride, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J61900.03
also known as J61900-03
Price (USD)/ Each
50.60
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
50.60
Add to cart
Phenoxybenzamine hydrochloride, Thermo Scientific Chemicals
Catalog numberJ61900.03
Price (USD)/ Each
50.60
-
Add to cart
Chemical Identifiers
CAS63-92-3
IUPAC Namehydrogen benzyl(2-chloroethyl)(1-phenoxypropan-2-yl)amine chloride
Molecular FormulaC18H23Cl2NO
InChI KeyVBCPVIWPDJVHAN-UHFFFAOYNA-N
SMILES[H+].[Cl-].CC(COC1=CC=CC=C1)N(CCCl)CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
FormPowder
Proton NMRConforms
Purity≥90.0%
Selective alpha adrenergic blocking agent. Irreversible calmodulin inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Selective alpha adrenergic blocking agent
  • Irreversible calmodulin inhibitor
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only