Thermo Scientific Chemicals

L-Methionine, Cell Culture Reagent

Catalog number: J61904.36
500 g, Each
Thermo Scientific Chemicals

L-Methionine, Cell Culture Reagent

Catalog number: J61904.36
500 g, Each
Quantity
Catalog number: J61904.36
also known as J61904-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
63-68-3
IUPAC Name
(2S)-2-amino-4-(methylsulfanyl)butanoic acid
Molecular Formula
C5H11NO2S
InChI Key
FFEARJCKVFRZRR-BYPYZUCNSA-N
SMILES
CSCC[C@H](N)C(O)=O
Assay from Supplier's CofA
≥99.0 to ≤ 101.0%
Comment
Endotoxin: ≤6.0EU/g
Impurity content
Iron, Fe: ≤10ppm
Optical Rotation
@ 20?C: +23.0 to +24.0? (c=2, 6N HCl, dry weight basis)
Heavy Metals (as Pb)
≤10ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text