Thermo Scientific Chemicals

Mycophenolic acid, 98%, Thermo Scientific Chemicals

Catalog number: J61905.03
1 g, Each
Thermo Scientific Chemicals

Mycophenolic acid, 98%, Thermo Scientific Chemicals

Catalog number: J61905.03
1 g, Each
Quantity
Catalog number: J61905.03
also known as J61905-03
Price (USD)
Quantity
-
Have Questions?

Description

It is used as a small molecule IMPDH inhibitor. mycophenolic Acid is used as an immunosuppressive agent with antiviral and antitumor effects. It potently inhibits inosine monophosphate dehydrogenase, thus inhibiting de novo GTP synthesis leading to decreased RNA and DNA synthesis. It also reversibly inhibits proliferation of T and B lymphocytes and antibody formation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a small molecule IMPDH inhibitor. mycophenolic Acid is used as an immunosuppressive agent with antiviral and antitumor effects. It potently inhibits inosine monophosphate dehydrogenase, thus inhibiting de novo GTP synthesis leading to decreased RNA and DNA synthesis. It also reversibly inhibits proliferation of T and B lymphocytes and antibody formation.

Solubility
Soluble in methanol at 50mg/ml

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from strong oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text