Mycophenolic acid, 98%, Thermo Scientific Chemicals
Mycophenolic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Mycophenolic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
1 g
5 g
Catalog number J61905.09
also known as J61905-09
Price (USD)/ Each
217.00
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
217.00
Add to cart
Mycophenolic acid, 98%, Thermo Scientific Chemicals
Catalog numberJ61905.09
Price (USD)/ Each
217.00
-
Add to cart
Specifications
Chemical Name or MaterialMycophenolic acid
CAS24280-93-1
Health Hazard 1H302-H341-H360D-H372
Health Hazard 2GHS H Statement
H302
Harmful if swallowed.
Health Hazard 3P201-P202-P260-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
View more
It is used as a small molecule IMPDH inhibitor. mycophenolic Acid is used as an immunosuppressive agent with antiviral and antitumor effects. It potently inhibits inosine monophosphate dehydrogenase, thus inhibiting de novo GTP synthesis leading to decreased RNA and DNA synthesis. It also reversibly inhibits proliferation of T and B lymphocytes and antibody formation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a small molecule IMPDH inhibitor. mycophenolic Acid is used as an immunosuppressive agent with antiviral and antitumor effects. It potently inhibits inosine monophosphate dehydrogenase, thus inhibiting de novo GTP synthesis leading to decreased RNA and DNA synthesis. It also reversibly inhibits proliferation of T and B lymphocytes and antibody formation.

Solubility
Soluble in methanol at 50mg/ml

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from strong oxidizing agent.
RUO – Research Use Only

General References:

  1. Eugui et al. Lymphocyte-selective cytostatic and immunosuppressive effects of mycophenolic acid in vitro: role of deoxyguanosine nucleotide depeletion. Scand.J.Immunol. 1991, 33, (2), 161-73.
  2. Allison et al. Mechanisms of action of mycophenolic acid. Ann.N.Y.Acad.Sci. 1993, 696, 63-87.