Thermo Scientific Chemicals

Luzindole, 97%, Thermo Scientific Chemicals

Catalog number: J61915.LB0
5 mg, Each
Thermo Scientific Chemicals

Luzindole, 97%, Thermo Scientific Chemicals

Catalog number: J61915.LB0
5 mg, Each
Quantity
Catalog number: J61915.LB0
also known as J61915-#0
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
117946-91-5
IUPAC Name
N-[2-(2-benzyl-1H-indol-3-yl)ethyl]acetamide
Molecular Formula
C19H20N2O
InChI Key
WVVXBPKOIZGVNS-UHFFFAOYSA-N
SMILES
CC(=O)NCCC1=C(CC2=CC=CC=C2)NC2=CC=CC=C12
Form
Powder or crystals or crystalline powder
Appearance (Color)
White to yellow
Assay from Supplier's CofA
≥96.0%

Description

Luzindole significantly antagonizes melatonin-mediated antinociception, suggesting that MEL-1B-R plays a further role as a mediator of melatonin modulation of pain signaling. It is used both in vitro and in vivo to evaluate the roles of melatonin receptor signaling in diverse pathways, including circadian rhythms, animal behavior, and melanophore response

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Luzindole significantly antagonizes melatonin-mediated antinociception, suggesting that MEL-1B-R plays a further role as a mediator of melatonin modulation of pain signaling. It is used both in vitro and in vivo to evaluate the roles of melatonin receptor signaling in diverse pathways, including circadian rhythms, animal behavior, and melanophore response

Solubility
Soluble in DMSO (100 mM), ethanol (100 mM), chloroform, methanol, and water.

Notes
Protect from heat. Incompatible with heat and oxidizing agents. Store at 4°C.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text