Phosphate-buffered saline (DPBS, 10X), Dulbecco's formula
Phosphate-buffered saline (DPBS, 10X), Dulbecco's formula
Thermo Scientific Chemicals
Phosphate-buffered saline (DPBS, 10X), Dulbecco's formula
Used as a buffer system to maintain cell culture media in the physiological pH range. In combination with glucose, it provides the principal energy sources of cell metabolism. It is involved to irrigate, wash and dilute mammalian cells.
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
2 L
Catalog number J61917.K2
also known as J61917-K2
Price (USD)
80.65
Special offer
Online exclusive
Ends: 26-Sep-2025
99.20
Save 18.55 (19%)
Each
-
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (USD)
80.65
Special offer
Online exclusive
Ends: 26-Sep-2025
99.20
Save 18.55 (19%)
Each
Add to cart
Specifications
Packaging TypePlastic bottle
Quantity1 L
FormLiquid
Unit SizeEach
Phosphate-buffered saline (DPBS, 10X), Dulbecco's formula is used as a buffer system to maintain cell culture media in the physiological pH range. In combination with glucose, it provides the principal energy sources of cell metabolism. It is involved to irrigate, wash and dilute mammalian cells. It plays an important role in the transportation of tissue samples and as diluents in various reagent preparations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
Ambient temperatures
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link