Thermo Scientific Chemicals

Phosphoenolpyruvic acid tricyclohexylammonium salt, Thermo Scientific Chemicals

Catalog number: J61943.03
1 g, Each
Thermo Scientific Chemicals

Phosphoenolpyruvic acid tricyclohexylammonium salt, Thermo Scientific Chemicals

Catalog number: J61943.03
1 g, Each
Quantity
Catalog number: J61943.03
also known as J61943-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
35556-70-8
IUPAC Name
tris(2-(phosphonooxy)prop-2-enoic acid); cyclohexanamine
Molecular Formula
C15H28NO18P3
InChI Key
WNMMIHPFMKMUPZ-UHFFFAOYSA-N
SMILES
NC1CCCCC1.OC(=O)C(=C)OP(O)(O)=O.OC(=O)C(=C)OP(O)(O)=O.OC(=O)C(=C)OP(O)(O)=O
Appearance (Color)
White
Water Content (Karl Fischer Titration)
6.0% max.
Form
Powder

Description

Metabolic intermediate in many kinase reactions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text