Thermo Scientific Chemicals

Benzotropine methanesulfonate, Thermo Scientific Chemicals

Catalog number: J61954.MC
100 mg, Each
Thermo Scientific Chemicals

Benzotropine methanesulfonate, Thermo Scientific Chemicals

Catalog number: J61954.MC
100 mg, Each
Quantity
Catalog number: J61954.MC
also known as J61954-MC
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
132-17-2
Appearance (Color)
White to off-white
Form
Powder
Assay (HPLC)
≥98.0%
Infrared spectrum
Conforms

Description

Benzotropine methanesulfonate is used to treat the symptoms of Parkinson?s disease and is currently in clincial trials for chronic back pain. It shows anti-muscarinic effects comparable to Atropine, and also demonstrates competitive anti-nicotinic action at the nAChR (nicotinic acetylcholine receptor).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water (81 mg/ml at 25°C), DMSO (50 mg/ml at 25°C), ethanol (81 mg/ml at 25°C), and ether (very slightly).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents. Store at 4°C.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text