Inosine-5'-monophosphate disodium salt hydrate
Inosine-5'-monophosphate disodium salt hydrate
Thermo Scientific Chemicals

Inosine-5'-monophosphate disodium salt hydrate

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number J61959.06
also known as J61959-06
Price (USD)/ Each
36.90
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
36.90
Add to cart
Inosine-5'-monophosphate disodium salt hydrate
Catalog numberJ61959.06
Price (USD)/ Each
36.90
-
Add to cart
Chemical Identifiers
CAS352195-40-5
IUPAC Namedisodium [(2R,3S,4R,5R)-3,4-dihydroxy-5-(6-oxo-6,9-dihydro-1H-purin-9-yl)oxolan-2-yl]methyl phosphate
Molecular FormulaC10H11N4Na2O8P
InChI KeyAANLCWYVVNBGEE-WCYUCLFNNA-L
SMILES[Na+].[Na+].O[C@@H]1[C@@H](COP([O-])([O-])=O)O[C@H]([C@@H]1O)N1C=NC2=C1N=CNC2=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline powder
Water Content (Karl Fischer Titration)≤27.0%
Assay from Suppliers CofA≥97.5%
Appearance (Color)White to pale cream
Heavy Metals≤10ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only