Chemical Identifiers
CAS352195-40-5
IUPAC Namedisodium [(2R,3S,4R,5R)-3,4-dihydroxy-5-(6-oxo-6,9-dihydro-1H-purin-9-yl)oxolan-2-yl]methyl phosphate
Molecular FormulaC10H11N4Na2O8P
InChI KeyAANLCWYVVNBGEE-WCYUCLFNNA-L
SMILES[Na+].[Na+].O[C@@H]1[C@@H](COP([O-])([O-])=O)O[C@H]([C@@H]1O)N1C=NC2=C1N=CNC2=O
View more