MES, 1.0M buffer soln., pH 5.0
MES, 1.0M buffer soln., pH 5.0
MES, 1.0M buffer soln., pH 5.0
MES, 1.0M buffer soln., pH 5.0
Thermo Scientific Chemicals

MES, 1.0M buffer soln., pH 5.0

CAS: 145224-94-8 | C6H15NO5S | 213.248 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
500 mL
Catalog number J61960.AK
also known as J61960-AK
Price (USD)
135.65
Special offer
Online exclusive
Ends: 26-Sep-2025
167.00
Save 31.35 (19%)
Each
-
Add to cart
Quantity:
250 mL
Request bulk or custom format
Price (USD)
135.65
Special offer
Online exclusive
Ends: 26-Sep-2025
167.00
Save 31.35 (19%)
Each
Add to cart
Specifications
CAS145224-94-8
Chemical Name or MaterialMES buffer soln.
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
Ask our AI about this Product
MES is used as a buffering agent in biology and biochemistry. It is utilized for resolving very small proteins on bis-tris gels. It is mainly used as a biological buffer in plant cell cultures. Buffer solutions are used as a means of keeping pH at a nearly constant value in a wide variety of chemical applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
MES is used as a buffering agent in biology and biochemistry. It is utilized for resolving very small proteins on bis-tris gels. It is mainly used as a biological buffer in plant cell cultures. Buffer solutions are used as a means of keeping pH at a nearly constant value in a wide variety of chemical applications.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Pagan, M.; Suazo, D.; del Toro, N.; Griebenow, K. A comparative study of different protein immobilization methods for the construction of an efficient nano-structured lactate oxidase-SWCNT-biosensor. Biosens. Bioelectron. 2015, 64, 138-146.
  2. Ojeda, I.; Barrejon, M.; Arellano, L. M.; Cortes, A. G.; Sedeno, P. Y., Langa, F.; Pingarrón, J. M. Grafted-double walled carbon nanotubes as electrochemical platforms for immobilization of antibodies using a metallic-complex chelating polymer: Application to the determination of adiponectin cytokine in serum. Biosens. Bioelectron. 2015, 74, 24-29.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link