Thermo Scientific Chemicals

Ciprofloxacin hydrochloride monohydrate, Thermo Scientific Chemicals

Catalog number: J61970.14
25 g, Each
Thermo Scientific Chemicals

Ciprofloxacin hydrochloride monohydrate, Thermo Scientific Chemicals

Catalog number: J61970.14
25 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J61970.14
also known as J61970-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
86393-32-0
Impurity content
Heavy metals: 0.002% max.
Form
Powder
Residue on ignition
0.1% max.
Water Content (Karl Fischer Titration)
4.7-6.7%
Appearance (Color)
White to off-white

Description

Ciprofloxacin hydrochloride has been suggested as a treatment for a wide range of infections caused by susceptible organisms including infections of urinary, respiratory, and gastrointestinal tracts, gonorrhoea and septicaemia. Nalidixic acid analogue.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ciprofloxacin hydrochloride has been suggested as a treatment for a wide range of infections caused by susceptible organisms including infections of urinary, respiratory, and gastrointestinal tracts, gonorrhoea and septicaemia. May be given by mouth or may be administered by intravenous infusion. Nalidixic acid analogue.

Solubility
Soluble in water (35mg/ml).

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text