trans-Zeatin, 97%, Thermo Scientific Chemicals
trans-Zeatin, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

trans-Zeatin, 97%, Thermo Scientific Chemicals

Plant growth hormone that stimulates cell division
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mg
100 mg
250 mg
Catalog number J61972.MF
also known as J61972-MF
Price (USD)/ Each
294.00
-
Add to cart
Quantity:
50 mg
Request bulk or custom format
Price (USD)/ Each
294.00
Add to cart
trans-Zeatin, 97%, Thermo Scientific Chemicals
Catalog numberJ61972.MF
Price (USD)/ Each
294.00
-
Add to cart
Chemical Identifiers
CAS1637-39-4
IUPAC Name(2E)-2-methyl-4-[(7H-purin-6-yl)amino]but-2-en-1-ol
Molecular FormulaC10H13N5O
InChI KeyUZKQTCBAMSWPJD-FARCUNLSSA-N
SMILESCC(CO)=C/CNC1=C2NC=NC2=NC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point206-214°C
Appearance (Color)White to off-white
Assay from Suppliers CofA≥96.0%
FormPowder or crystals or crystalline powder
trans-Zeatin regulates cell division, development, and nutrient processing. It acts as a high-affinity ligand for a number of receptors, including arabidopsis histidine kinases (AHKs), specifically AHK3, which is important in mediating plant development.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-Zeatin regulates cell division, development, and nutrient processing. It acts as a high-affinity ligand for a number of receptors, including arabidopsis histidine kinases (AHKs), specifically AHK3, which is important in mediating plant development.

Solubility
Soluble in DMSO and DMF

Notes
Store at -20°C. Incompatible with heat and oxidizing agents.
RUO – Research Use Only

General References:

  1. Iwona Żur; Ewa Dubas; Monika Krzewska; Piotr Waligórski; Michał Dziurka; Franciszek Janowiak. Hormonal requirements for effective induction of microspore embryogenesis in triticale (x Triticosecale Wittm.) anther cultures. Plant Cell Reports. 2015, 34, (1), 47-62.
  2. Robbin L Koenig; Roy O Morris; Joe C Polacco. tRNA is the source of low-level trans-zeatin production in Methylobacterium spp. Journal of Bacteriology. 2002, 184, (7), 1832-1842.