Thermo Scientific Chemicals

Nickel(II) sulfate, 0.5M aq. soln., Thermo Scientific Chemicals

Catalog number: J61993.AE
100 mL, Each
Thermo Scientific Chemicals

Nickel(II) sulfate, 0.5M aq. soln., Thermo Scientific Chemicals

Catalog number: J61993.AE
100 mL, Each
Quantity
Catalog number: J61993.AE
also known as J61993-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10101-97-0
IUPAC Name
nickel(2+) hexahydrate sulfate
Molecular Formula
H12NiO10S
InChI Key
RRIWRJBSCGCBID-UHFFFAOYSA-L
SMILES
O.O.O.O.O.O.[Ni++].[O-]S([O-])(=O)=O
Appearance (Color)
Green
Composition
Nickel (II) Sulfate: 500 mM
Form
Liquid

Description

Nickel(II) sulfate, 0.5M aq. soln. is used in nickel plating, as mordant in dyeing and in printing textiles. Columns used in polyhistidine-tagging, useful in biochemistry and molecular biology, are regenerated with nickel sulfate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nickel(II) sulfate, 0.5M aq. soln. is used in nickel plating, as mordant in dyeing and in printing textiles. Columns used in polyhistidine-tagging, useful in biochemistry and molecular biology, are regenerated with nickel sulfate.

Solubility
Fully miscible in water.

Notes
Protect from heat.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text