Thermo Scientific Chemicals

Fluconazole, 99%, Thermo Scientific Chemicals

Catalog number: J62015.14
25 g, Each
Thermo Scientific Chemicals

Fluconazole, 99%, Thermo Scientific Chemicals

Catalog number: J62015.14
25 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J62015.14
also known as J62015-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
86386-73-4
IUPAC Name
2-(2,4-difluorophenyl)-1,3-bis(1H-1,2,4-triazol-1-yl)propan-2-ol
Molecular Formula
C13H12F2N6O
InChI Key
RFHAOTPXVQNOHP-UHFFFAOYSA-N
SMILES
OC(CN1C=NC=N1)(CN1C=NC=N1)C1=CC=C(F)C=C1F
Form
Powder
Appearance (Color)
White to pale cream
Assay (special)
≥99.0%

Description

Fluconazole is used to treat a variety of fungal infections, especially Candida infections of the vagina (yeast infections), mouth, throat and bloodstream. It is also used by the people having weak immune system such as persons with neutropenia due to cancer chemotherapy, transplanted patients and premature infants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description
• Fluconazole, 98% is a triazole compound showing antifungal activity mainly against Candida, Coccidioides, and Cryptococcus species
• It has been shown to inhibit fungal cytochrome P-450 sterol C-14 α-demethylation, resulting in the accumulation of fungal 14 α-methyl sterols, the loss of normal fungal sterols, and associated fungistatic activity
• It also inhibits cytochrome P450 family 2 subfamily c member 9 (CYP2C9)
• Mammalian cell demethylation is much less susceptible to fluconazole inhibition
Applications
• Fluconazole has fungistatic activity against many fungal strains including Candida albicans, Candida glabrata, Candida parapsilosis, Candida tropicalis, and Cryptococcus neoformans.
• Fluconazole interferes with fungal ergosterol synthesis and downregulates the metallothionein gene
• In biological experiments, this compound has been used to assess minimum inhibitory concentration in (MIC)-fungal assays
• It has also been used as a positive control in antifungal assay of plants
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text