Glimepiride, Thermo Scientific Chemicals
Glimepiride, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Glimepiride, Thermo Scientific Chemicals

A sulfonylurea hypoglecemic agent and potent potassium channel blocker
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
100 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J62032.03
also known as J62032-03
Price (USD)/ Each
390.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
390.00
Add to cart
Glimepiride, Thermo Scientific Chemicals
Catalog numberJ62032.03
Price (USD)/ Each
390.00
-
Add to cart
Chemical Identifiers
CAS93479-97-1
IUPAC Name3-ethyl-4-methyl-N-{2-[4-({[(4-methylcyclohexyl)carbamoyl]amino}sulfonyl)phenyl]ethyl}-2-oxo-2,5-dihydro-1H-pyrrole-1-carboxamide
Molecular FormulaC24H34N4O5S
InChI KeyWIGIZIANZCJQQY-UHFFFAOYSA-N
SMILESCCC1=C(C)CN(C(=O)NCCC2=CC=C(C=C2)S(=O)(=O)NC(=O)NC2CCC(C)CC2)C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay from Suppliers CofA≥95.0%
Glimepiride induces the PI3 kinase (PI3K) and Akt pathway, along with insulin receptor substrate-1/2 and endothelial nitric oxide synthase. Glimepiride also increases osteoblast proliferation and differentiation, which is thought to be related to its ability to activate the PI3K and Akt pathway. Furthermore, Glimepiride enhances intrinsic peroxisome proliferator-activated receptor γ activity. Glimepiride also increases protein expression of glucose transports 1 and 4, and is a potent KIR channel blocker. Potent Kir6 (KATP) channel blocker and anti-diabetic agent. Inhibits pinacidil-activated cardiac Kir6 channels with an IC50 of 6.8 nM.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glimepiride induces the PI3 kinase (PI3K) and Akt pathway, along with insulin receptor substrate-1/2 and endothelial nitric oxide synthase. Glimepiride also increases osteoblast proliferation and differentiation, which is thought to be related to its ability to activate the PI3K and Akt pathway. Furthermore, Glimepiride enhances intrinsic peroxisome proliferator-activated receptor γ activity. Glimepiride also increases protein expression of glucose transports 1 and 4, and is a potent KIR channel blocker. Potent Kir6 (KATP) channel blocker and anti-diabetic agent. Inhibits pinacidil-activated cardiac Kir6 channels with an IC50 of 6.8 nM.

Solubility
Soluble in DMSO

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Lawrence et al. Effect of metabolic inhibition on glimepiride block of native and cloned cardiac sacrolemmal KATP channels. Br.J.Pharmacol. 2002, 136, (5), 746-52.
  2. SE Nissen.; SJ Nicholls.; K Wolski.; R Nesto.; S Kupfer. Comparison of pioglitazone vs glimepiride on progression of coronary atherosclerosis in patients with type 2 diabetes: the periscope randomized controlled . JAMA. 2008, 299, (13), 1561-1573.