Thermo Scientific Chemicals

Phosphate-buffered saline (PBS, 10X), pH 7.4

Catalog number: J62036.K3
2 L, Each
Thermo Scientific Chemicals

Phosphate-buffered saline (PBS, 10X), pH 7.4

Catalog number: J62036.K3
2 L, Each
Capacity
Catalog number: J62036.K3
also known as J62036-K3
Price (USD)
Quantity
-
Have Questions?
Form
Liquid
Comment
All components are dissolved in 18.2 megohm UV treated water, adjusted pH and filtered with sub-micron filter
Appearance (Color)
Clear colorless
pH
7.4 ± 0.2, after 1X dilution with DI water
Composition
1.37 M Sodium Chloride, 27 mM KCl, 101.4 mM Phosphate Buffer, 17.6 Potassium Phosphate Monobasic

Description

Phosphate-buffered saline (PBS, 10X) is an isotonic, non-toxic to most cells and is used in biological research. It is associated with ethylenediaminetetraacetic acid (EDTA) and is used to disengage attached and clumped cells. It also helps to maintain a constant pH.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text