Aconitine, 98%, Thermo Scientific Chemicals
Aconitine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Aconitine, 98%, Thermo Scientific Chemicals

Have Questions?
Quantity:
250 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J62056.MD
also known as J62056-MD
Price (USD)/ Each
872.00
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
872.00
Add to cart
Aconitine, 98%, Thermo Scientific Chemicals
Catalog numberJ62056.MD
Price (USD)/ Each
872.00
-
Add to cart
Chemical Identifiers
CAS302-27-2
IUPAC Name(1S,2R,3R,4R,5R,6S,7S,8R,9R,13R,14R,16S,17S,18R)-8-(acetyloxy)-11-ethyl-5,7,14-trihydroxy-6,16,18-trimethoxy-13-(methoxymethyl)-11-azahexacyclo[7.7.2.1²,⁵.0¹,¹⁰.0³,⁸.0¹³,¹⁷]nonadecan-4-yl benzoate
Molecular FormulaC34H47NO11
InChI KeyXFSBVAOIAHNAPC-XTHSEXKGSA-N
SMILESCCN1C[C@@]2(COC)[C@H]3[C@@H](OC)[C@H]4C1[C@@]3([C@@H]1C[C@@]3(O)[C@H](OC(=O)C5=CC=CC=C5)[C@@H]1[C@]4(OC(C)=O)[C@@H](O)[C@@H]3OC)[C@H](C[C@H]2O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Assay (HPLC)97.5% min.
FormPowder
Opens tetrodotoxin-sensitive sodium channels. Useful for creating models of cardiac arrhythmia

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Opens tetrodotoxin-sensitive sodium channels
  • Useful for creating models of cardiac arrhythmia
RUO – Research Use Only