Thermo Scientific Chemicals
Diosmin, Thermo Scientific Chemicals
Catalog number: J62073.14
25 g, Each
Thermo Scientific Chemicals
Diosmin, Thermo Scientific Chemicals
Catalog number: J62073.14
25 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J62073.14
also known as J62073-14
Price (USD)
Quantity
-
Have Questions?
Description
Diosmin exhibits anti-proliferative and anti-inflammatory. It is used in the treatment of venous disease, i.e., chronic venous insufficiency (CVI) and hemorrhoidal disease (HD).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in DMSO (50 mg/ml), water (122 mg/ml at 25°C), and ethanol (<1mg/mL at 25°C).
Notes
Protect from heat. Incompatible with heat and oxidizing agents. Store at 4°C.