Retinol, 95%, synthetic, Thermo Scientific Chemicals
Retinol, 95%, synthetic, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Retinol, 95%, synthetic, Thermo Scientific Chemicals

All-trans-retinol, Vitamin A, retinol, CAS # 68-26-8, is a fat-soluble vitamin that binds retinoid receptors within cells.
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
100 mg
250 mg
500 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J62079.03
also known as J62079-03
Price (USD)/ Each
270.65
Online exclusive
300.00 
Save 29.35 (10%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
270.65
Online exclusive
300.00 
Save 29.35 (10%)
Add to cart
Retinol, 95%, synthetic, Thermo Scientific Chemicals
Catalog numberJ62079.03
Price (USD)/ Each
270.65
Online exclusive
300.00 
Save 29.35 (10%)
-
Add to cart
Chemical Identifiers
CAS68-26-8
IUPAC Name(2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraen-1-ol
Molecular FormulaC20H30O
InChI KeyFPIPGXGPPPQFEQ-OVSJKPMPSA-N
SMILESC\C(=C/CO)\C=C\C=C(/C)\C=C\C1=C(C)CCCC1(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to orange
FormPowder
Assay (special)≥94%
Melting Point60 to 65°C
Proton NMRConforms to structure
It induces cell differentiation and apoptosis of some cancer cell types and inhibiting carcinogenesis. It plays an essential role in metabolic functioning of the retina, the growth of and differentiation of epithelial tissue, the growth of bone, reproduction, and the immune response. It acts as a ligand for both the retinoic acid receptor (RAR) and the retinoid X receptor (RXR) that act as transcription factors to regulate the growth and differentiation of normal and malignant cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description
• All-trans-retinol, commonly known as Vitamin A, is a fat-soluble essential nutrient
• All-trans-retinol is involved in hormonal signaling via the retinoid receptors
• It is an important regulator of cell division and apoptosis
• Deficiency can lead to vision, bone, immune, and skin disorders

Applications
• Suitable for research on cell differentiation, embryonic development, immunology and more
• Laboratory grade quality, for research use only

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. Stephensen, C.B.; Lietz, G. Vitamin A in resistance to and recovery from infection: relevance to SARS-CoV2. Br J Nutr. 2021, 126(11), 1663-1672.
  2. He, J.; Gu, Y.; Zhang, S. Vitamin A and Breast Cancer Survival: A Systematic Review and Meta-analysis. Clin Breast Cancer. 2018, 18(6), e1389-e1400.