Thermo Scientific Chemicals

Retinol, 95%, synthetic, Thermo Scientific Chemicals

Catalog number: J62079.MD
250 mg, Each
Thermo Scientific Chemicals

Retinol, 95%, synthetic, Thermo Scientific Chemicals

Catalog number: J62079.MD
250 mg, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J62079.MD
also known as J62079-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
68-26-8
IUPAC Name
(2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraen-1-ol
Molecular Formula
C20H30O
InChI Key
FPIPGXGPPPQFEQ-OVSJKPMPSA-N
SMILES
C\C(=C/CO)\C=C\C=C(/C)\C=C\C1=C(C)CCCC1(C)C
Appearance (Color)
Yellow to orange
Form
Powder
Assay (special)
≥94%
Melting Point
60 to 65°C
Proton NMR
Conforms to structure

Description

It induces cell differentiation and apoptosis of some cancer cell types and inhibiting carcinogenesis. It plays an essential role in metabolic functioning of the retina, the growth of and differentiation of epithelial tissue, the growth of bone, reproduction, and the immune response. It acts as a ligand for both the retinoic acid receptor (RAR) and the retinoid X receptor (RXR) that act as transcription factors to regulate the growth and differentiation of normal and malignant cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description
• All-trans-retinol, commonly known as Vitamin A, is a fat-soluble essential nutrient
• All-trans-retinol is involved in hormonal signaling via the retinoid receptors
• It is an important regulator of cell division and apoptosis
• Deficiency can lead to vision, bone, immune, and skin disorders

Applications
• Suitable for research on cell differentiation, embryonic development, immunology and more
• Laboratory grade quality, for research use only

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text