TRIS, 1.0M buffer soln., pH 9.0 is used as buffers for stabilizing and storing nucleic acids and proteins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
TRIS, 1.0M buffer soln., pH 9.0 is used as buffers for stabilizing and storing nucleic acids and proteins.
Solubility
Soluble in water and organic solvents.
Notes
Store in cool place. Keep container
RUO – Research Use Only
DE Soltis.; CH Haufler.; DC Darrow.; GJ Gastony. Starch gel electrophoresis of ferns: a compilation of grinding buffers, gel and electrodebuffers, and staining schedules. American fern journal. 1983, 73 ,(1),9-27.
DE Soltis.; CH Haufler.; DC Darrow.; GJ Gastony. Starch gel electrophoresis of ferns: a compilation of grinding buffers, gel and electrodebuffers, and staining schedules. J. Appl. Cryst.1991, 24 ,409-411.