beta-Nicotinamide adenine dinucleotide phosphate reduced tetrasodium salt, 95%, Thermo Scientific Chemicals
beta-Nicotinamide adenine dinucleotide phosphate reduced tetrasodium salt, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

beta-Nicotinamide adenine dinucleotide phosphate reduced tetrasodium salt, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
500 mg
Catalog number J62089.MC
also known as J62089-MC
Price (USD)/ Each
200.65
Online exclusive
223.00 
Save 22.35 (10%)
-
Add to cart
Quantity:
100 mg
Request bulk or custom format
Price (USD)/ Each
200.65
Online exclusive
223.00 
Save 22.35 (10%)
Add to cart
beta-Nicotinamide adenine dinucleotide phosphate reduced tetrasodium salt, 95%, Thermo Scientific Chemicals
Catalog numberJ62089.MC
Price (USD)/ Each
200.65
Online exclusive
223.00 
Save 22.35 (10%)
-
Add to cart
Chemical Identifiers
CAS2646-71-1
IUPAC Nametetrasodium [(2R,3R,4R,5R)-5-(6-amino-9H-purin-9-yl)-3-hydroxy-4-(phosphonatooxy)oxolan-2-yl]methyl ({[(2R,3S,4R,5R)-5-(3-carbamoyl-1,4-dihydropyridin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)phosphonate
Molecular FormulaC21H26N7Na4O17P3
InChI KeyWYWWVJHQDVCHKF-NRTBITFTNA-J
SMILES[Na+].[Na+].[Na+].[Na+].NC(=O)C1=CN(C=CC1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](OP([O-])([O-])=O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light cream to yellow
FormCrystalline powder
Assay (HPLC)≥ 95.0%
Water Content (Karl Fischer Titration)≤ 8.0%
Solution TestClear solution (50 mg/ml in water)
NADPH tetra sodium salt is used as an ubiquitous cofactor and biological reducing agent. β-NADPH is a coenzyme found in all living cells and is involved in redox reactions carrying electrons from one reaction to another. It is used as an electron donor, cofactor for many redox enzymes including nitric oxide synthetase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
NADPH tetra sodium salt is used as an ubiquitous cofactor and biological reducing agent. β-NADPH is a coenzyme found in all living cells and is involved in redox reactions carrying electrons from one reaction to another. It is used as an electron donor, cofactor for many redox enzymes including nitric oxide synthetase.

Solubility
Soluble in water (50 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature -20°C. Light sensitive. Keep away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. D Gallego.; V Gil.; J Aleu. Pharmacological characterization of purinergic inhibitory neuromuscular transmission in the human colon. Neurogastroenterology & Motility. 2011, 23, (8), 792-e338.
  2. V Gil.; M Martínez-Cutillas.; N Mane. P2Y1 knockout mice lack purinergic neuromuscular transmission in the antrum and cecum. Neurogastroenterology & Motility. 2013, 25, (3), e170-e182.