Thermo Scientific Chemicals

beta-Nicotinamide adenine dinucleotide phosphate reduced tetrasodium salt, 95%, Thermo Scientific Chemicals

Catalog number: J62089.ME
500 mg, Each
Thermo Scientific Chemicals

beta-Nicotinamide adenine dinucleotide phosphate reduced tetrasodium salt, 95%, Thermo Scientific Chemicals

Catalog number: J62089.ME
500 mg, Each
Quantity
Catalog number: J62089.ME
also known as J62089-ME
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2646-71-1
IUPAC Name
tetrasodium [(2R,3R,4R,5R)-5-(6-amino-9H-purin-9-yl)-3-hydroxy-4-(phosphonatooxy)oxolan-2-yl]methyl ({[(2R,3S,4R,5R)-5-(3-carbamoyl-1,4-dihydropyridin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)phosphonate
Molecular Formula
C21H26N7Na4O17P3
InChI Key
WYWWVJHQDVCHKF-NRTBITFTNA-J
SMILES
[Na+].[Na+].[Na+].[Na+].NC(=O)C1=CN(C=CC1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](OP([O-])([O-])=O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O
Appearance (Color)
Light cream to yellow
Form
Crystalline powder
Assay (HPLC)
≥ 95.0%
Water Content (Karl Fischer Titration)
≤ 8.0%
Solution Test
Clear solution (50 mg/ml in water)

Description

NADPH tetra sodium salt is used as an ubiquitous cofactor and biological reducing agent. β-NADPH is a coenzyme found in all living cells and is involved in redox reactions carrying electrons from one reaction to another. It is used as an electron donor, cofactor for many redox enzymes including nitric oxide synthetase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
NADPH tetra sodium salt is used as an ubiquitous cofactor and biological reducing agent. β-NADPH is a coenzyme found in all living cells and is involved in redox reactions carrying electrons from one reaction to another. It is used as an electron donor, cofactor for many redox enzymes including nitric oxide synthetase.

Solubility
Soluble in water (50 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature -20°C. Light sensitive. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text