L-Lysine monohydrochloride, Cell Culture Reagent
L-Lysine monohydrochloride, Cell Culture Reagent
Thermo Scientific Chemicals

L-Lysine monohydrochloride, Cell Culture Reagent

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
1 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J62099.36
also known as J62099-36
Price (USD)/ Each
117.00
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
117.00
Add to cart
L-Lysine monohydrochloride, Cell Culture Reagent
Catalog numberJ62099.36
Price (USD)/ Each
117.00
-
Add to cart
Chemical Identifiers
CAS657-27-2
IUPAC Name(2S)-2,6-diaminohexanoic acid hydrochloride
Molecular FormulaC6H15ClN2O2
InChI KeyBVHLGVCQOALMSV-JEDNCBNOSA-N
SMILESCl.NCCCC[C@H](N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Heavy Metals (as Pb)≤10ppm
CommentEndotoxins: ≤6.0EU/g
Impurity contentArsenic as AS2O3: ≤1ppm
Impurity contentIron, Fe: ≤10ppm
Impurity contentSulphate, SO4: ≤0.02% (≤200ppm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only