Thermo Scientific Chemicals

Acrylamide (30%) Solutions

Catalog number: J62100.K2
1 L, Each
Thermo Scientific Chemicals

Acrylamide (30%) Solutions

Catalog number: J62100.K2
1 L, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J62100.K2
also known as J62100-K2
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
79-06-1
IUPAC Name
prop-2-enamide
Molecular Formula
C3H5NO
InChI Key
HRPVXLWXLXDGHG-UHFFFAOYSA-N
SMILES
NC(=O)C=C
Appearance (Color)
Clear
Form
Liquid
Comment
Composition: Acrylamide, 30% concentration

Description

Acrylamide is a commonly used matrix for electrophoretic separation of nucleic acids and proteins. Acrylamide is used as a chemical intermediate in production of polyacrylamides, for use in protein electrophoresis (PAGE), synthesis of dyes and copolymers for contact lenses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acrylamide is a commonly used matrix for electrophoretic separation of nucleic acids and proteins. Acrylamide is used as a chemical intermediate in production of polyacrylamides, for use in protein electrophoresis (PAGE), synthesis of dyes and copolymers for contact lenses.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Light sensitive. Keep away from Acids, Oxidizing agents, Iron and iron salts., Copper, Brass, Free radical initiators.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text