Informational:Informational:This product is animal origin and can only be shipped within the United States
Thermo Scientific Chemicals

Glutamic pyruvic transaminase, porcine heart

Catalog number: J62109.4I
1 kU, Each
Thermo Scientific Chemicals

Glutamic pyruvic transaminase, porcine heart

Catalog number: J62109.4I
1 kU, Each
Quantity
Catalog number: J62109.4I
also known as J62109-4I
Price (USD)
Have Questions?

Chemical Identifiers

CAS
9000-86-6
IUPAC Name
1-[4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]-5-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular Formula
C11H16N2O6
InChI Key
VVJYUAYZJAKGRQ-UHFFFAOYNA-N
SMILES
CC1=CN(C2CC(O)C(O)C(CO)O2)C(=O)NC1=O
Assay (unspecified)
Glyceraldehyde-3-P-Dehydrogenase: 0.001% max.
Assay (unspecified)
Lactate dehydrogrenase: 0.005% max.
Assay (unspecified)
Protein content (biuret): 40% min.
Unit definition
One unit converts one micromole of alpha-ketoglutarate to L-glutamate per minute at pH 7.5 and 37?C
Assay (unspecified)
Activity: 100 units/mg

Description

It is used for research and clinical diagnostic assays for liver disease. Alanine transaminase (GPT/ALT) is used in multi-analyte clinical chemistry controls and calibrators for major IVD manufacturers. The alanine aminotransferase (ALT) blood test is typically used to detect liver injury. Some of the other applications include Diagnostic Controls, calibrators & standards, immunoassays, clinical chemistry, testing/assay validation, life Science, manufacturing, validation Studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text