Thermo Scientific Chemicals

beta-Glycerophosphate, 200mM soln., Thermo Scientific Chemicals

Catalog number: J62121.AE
100 mL, Each
Thermo Scientific Chemicals

beta-Glycerophosphate, 200mM soln., Thermo Scientific Chemicals

Catalog number: J62121.AE
100 mL, Each
Quantity
Catalog number: J62121.AE
also known as J62121-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
819-83-0
IUPAC Name
tetrasodium 3-(phosphonatooxy)pentane-1,5-diol 3-(phosphonatooxy)propane-1,2-diol
Molecular Formula
C8H18Na4O12P2
InChI Key
WENPEHSLKZUZKX-UHFFFAOYNA-J
SMILES
[Na+].[Na+].[Na+].[Na+].OCC(O)COP([O-])([O-])=O.OCCC(CCO)OP([O-])([O-])=O
Comment
Component is dissolved in UV treated 18.2 megohm-cm ± 1 water, and filtered through 0.22 micron filter.
Appearance (Color)
Clear colorless
Form
Liquid

Description

β-Glycerolphosphate (Glycerol-2-Phosphate) is used in the development of hydrogels and scaffolds that have applications in tissue engineering and cell growth and differentiation. It has also been used as an additive in isolation mediums, similarly providing available phosphate to the isolate. It is a phosphate group donor in matrix mineralization studies that acts as a protein phosphatase inhibitor. Accelerates calcification in vascular smooth muscle cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
β-Glycerolphosphate (Glycerol-2-Phosphate) is used in the development of hydrogels and scaffolds that have applications in tissue engineering and cell growth and differentiation. It has also been used as an additive in isolation mediums, similarly providing available phosphate to the isolate. It is a phosphate group donor in matrix mineralization studies that acts as a protein phosphatase inhibitor. Accelerates calcification in vascular smooth muscle cells.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text