3,3',5,5'-Tetramethylbenzidine soln., Ready-to-Use, precipitating, high sensitivity, Thermo Scientific Chemicals
3,3',5,5'-Tetramethylbenzidine soln., Ready-to-Use, precipitating, high sensitivity, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,3',5,5'-Tetramethylbenzidine soln., Ready-to-Use, precipitating, high sensitivity, Thermo Scientific Chemicals

Have Questions?
Quantity:
500 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J62143.AP
also known as J62143-AP
Price (USD)/ Each
189.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
223.00 
Save 33.35 (15%)
-
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)/ Each
189.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
223.00 
Save 33.35 (15%)
Add to cart
3,3',5,5'-Tetramethylbenzidine soln., Ready-to-Use, precipitating, high sensitivity, Thermo Scientific Chemicals
Catalog numberJ62143.AP
Price (USD)/ Each
189.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
223.00 
Save 33.35 (15%)
-
Add to cart
Chemical Identifiers
CAS54827-17-7
IUPAC Name3,3',5,5'-tetramethyl-[1,1'-biphenyl]-4,4'-diamine
Molecular FormulaC16H20N2
InChI KeyUAIUNKRWKOVEES-UHFFFAOYSA-N
SMILESCC1=CC(=CC(C)=C1N)C1=CC(C)=C(N)C(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless
Assay from Suppliers CofADot Blot: Passes Test
FormLiquid
ClarityTransparent
Excellent colorimetric substrate for detection of horseradish peroxidase labelled probes and is used with peroxidase and peroxidase coupled systems, particularly in ELISA techniques, blue result

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Excellent colorimetric substrate for detection of horseradish peroxidase labelled probes and is used with peroxidase and peroxidase-coupled systems, particularly in ELISA techniques, blue result
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only