Leuprolide
Thermo Scientific Chemicals

Leuprolide

For cellular and molecular biology applications
Have Questions?
Quantity:
5 mg
Catalog number J62147.LB0
also known as J62147-#0
Price (USD)/ Each
188.00
-
Add to cart
Quantity:
5 mg
Request bulk or custom format
Price (USD)/ Each
188.00
Add to cart
Leuprolide
Catalog numberJ62147.LB0
Price (USD)/ Each
188.00
-
Add to cart
Specifications
Purification MethodHPLC
Common NameLeuprorelin
FormatPowder
For Use With (Application)Highly active luteinizing hormone releasing hormone (LHRH) agonist
Molecular Weight (g/mol)1209.41
View more
Highly active luteinizing hormone releasing hormone (LHRH) agonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only