Thermo Scientific Chemicals

Aloin, Thermo Scientific Chemicals

Catalog number: J62153.MC
100 mg, Each
Thermo Scientific Chemicals

Aloin, Thermo Scientific Chemicals

Catalog number: J62153.MC
100 mg, Each
Quantity
Catalog number: J62153.MC
also known as J62153-MC
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1415-73-2
IUPAC Name
(10R)-1,8-dihydroxy-3-(hydroxymethyl)-10-[(2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]-9,10-dihydroanthracen-9-one
Molecular Formula
C21H22O9
InChI Key
AFHJQYHRLPMKHU-WEZNYRQKSA-N
SMILES
OC[C@H]1O[C@H]([C@H](O)[C@@H](O)[C@@H]1O)[C@@H]1C2=CC=CC(O)=C2C(=O)C2=C(O)C=C(CO)C=C12
Appearance (Color)
Yellow
Form
Powder
Purity
98.20% (HPLC)

Description

Aloe exudant, used as a stimulant-laxative

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text