Thermo Scientific Chemicals

Calciferol, 97%, Thermo Scientific Chemicals

Catalog number: J62163.06
5g, Each
Thermo Scientific Chemicals

Calciferol, 97%, Thermo Scientific Chemicals

Catalog number: J62163.06
5g, Each
Quantity
Catalog number: J62163.06
also known as J62163-06
Price (USD)
Have Questions?

Chemical Identifiers

CAS
50-14-6
Optical Rotation
+103 to + 106º
Assay (HPLC)
97.0 to 103.0 %
Identification (FTIR)
Conforms to structure
Form
Crystalline powder
Assay (unspecified)
Reducing substances: ≤ 20 ppm

Description

Calciferol has a strong inhibitory effect against bladder tumor promotion by sodium saccharin and induces cell differentiation in leukemia cells. Calciferol has also been shown to be an inhibitor of DNA polymerase. Calciferol with calcium is used to treat or prevent bone loss (osteoporosis). Calciferol is also used with other medications to treat low levels of calcium or phosphate caused by certain disorders (such as hypoparathyroidism, pseudohypoparathyroidism, familial hypophosphatemia). It is often used in kidney disease to keep calcium levels normal and allow normal bone growth.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in ethanol (80 mg/ml at 25°C), water (<1mg/mL at 25°C), methanol, DMF (∼20 mg/ml), and DMSO (20 mg/ml at 25°C).

Notes
Store at 4°C. Incompatible with heat and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text