Thermo Scientific Chemicals

L-Glutathione, reduced, 98+%, Thermo Scientific Chemicals

Catalog number: J62166.14
25 g, Each
Thermo Scientific Chemicals

L-Glutathione, reduced, 98+%, Thermo Scientific Chemicals

Catalog number: J62166.14
25 g, Each
Quantity
Catalog number: J62166.14
also known as J62166-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
70-18-8
IUPAC Name
2-amino-4-({1-[(carboxymethyl)carbamoyl]-2-sulfanylethyl}carbamoyl)butanoic acid
Molecular Formula
C10H17N3O6S
InChI Key
RWSXRVCMGQZWBV-UHFFFAOYNA-N
SMILES
NC(CCC(=O)NC(CS)C(=O)NCC(O)=O)C(O)=O
Optical Rotation
-15.5 to -17.5°
Appearance (Color)
White
Form
Crystalline powder
Assay from Supplier's CofA
≥98.0%
Heavy Metals
≤10 ppm

Description

A tripeptide used for the reductive cleavage of disulfide bonds L-Glutathione is used in the treatment of lung diseases for patients who are HIV positive. It protects the cancerous cells by conferring resistance to chemotherapeutic drugs. It is involved in many aspects of metabolism including transport of g-glutanyl amino acids and reductive cleavage of disulfide bonds. As an antioxidant, it prevents damage to important cellular components that arise due to reactive oxygen species like free radicals and peroxide. It is also used to decrease the concentrations of inflammatory cytokines (IL-6, IL-18) as well as involved in increasing the level of serum Ca2+ ions. It is also used in white wine production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water, dimethyl formamide, ethanol, phosphate buffered saline, diluted alcohol, liquid ammonia and dimethyl sulfoxide.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text