Thermo Scientific Chemicals

Amiloride hydrochloride dihydrate, Thermo Scientific Chemicals

Catalog number: J62168.03
1 g, Each
Thermo Scientific Chemicals

Amiloride hydrochloride dihydrate, Thermo Scientific Chemicals

Catalog number: J62168.03
1 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J62168.03
also known as J62168-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
17440-83-4
IUPAC Name
hydrogen 3,5-diamino-6-chloro-N-(diaminomethylidene)pyrazine-2-carboxamide dihydrate chloride
Molecular Formula
C6H13Cl2N7O3
InChI Key
LTKVFMLMEYCWMK-UHFFFAOYSA-N
SMILES
[H+].O.O.[Cl-].NC(N)=NC(=O)C1=NC(Cl)=C(N)N=C1N
Assay (HPLC)
98.0% min.
Water Content (Karl Fischer Titration)
16.9% max.

Description

Amiloride hydrochloride dihydrate is used as a calcium channel and sodium channel protein inhibitor. It reduces electrical potential across tubular epithelium, inhibits angiogenesis and inhibits capillary morphogenesis completely and reversibly at approximately 130 ?M. Amiloride
  • HCl is an inhibitor of Sodium/Potassium-ATPase Protein. Defines the I2A-amiloride sensitive and I2B-amiloride insensitive imidazoline binding Blocks TRPP3, acid sensing- (ASIC) and mechanogated membrane-ion channels, as well as the Na+/H+ exchanger. Also inhibits urokinase-type plasminogen activator (uPA); has no effect on tissue-type plasminogen activator.

    This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

    Applications
    Amiloride hydrochloride dihydrate is used as a calcium channel and sodium channel protein inhibitor. It reduces electrical potential across tubular epithelium, inhibits angiogenesis and inhibits capillary morphogenesis completely and reversibly at approximately 130 μM. Amiloride • HCl is an inhibitor of Sodium/Potassium-ATPase Protein. Defines the I2A-amiloride sensitive and I2B-amiloride insensitive imidazoline binding Blocks TRPP3, acid sensing- (ASIC) and mechanogated membrane-ion channels, as well as the Na+/H+ exchanger. Also inhibits urokinase-type plasminogen activator (uPA); has no effect on tissue-type plasminogen activator.

    Solubility
    Soluble in water at 50mg/ml

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
    RUO – Research Use Only

    Figures

    Documents & Downloads

    Certificates

      Frequently asked questions (FAQs)

      Citations & References

      Search citations by name, author, journal title or abstract text