Thermo Scientific Chemicals

Adenosine-3',5'-cyclic monophosphate sodium salt, 99%

Catalog number: J62174.03
1 g, Each
Thermo Scientific Chemicals

Adenosine-3',5'-cyclic monophosphate sodium salt, 99%

Catalog number: J62174.03
1 g, Each
Quantity
Catalog number: J62174.03
also known as J62174-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
37839-81-9
IUPAC Name
sodium (4aR,6R,7R,7aS)-6-(6-amino-9H-purin-9-yl)-7-hydroxy-2-oxo-hexahydro-2λ⁵-furo[3,2-d][1,3,2]dioxaphosphinin-2-olate
Molecular Formula
C10H11N5NaO6P
InChI Key
BXJBFCKTIWRKMQ-GWKNMROSNA-M
SMILES
[Na+].NC1=C2N=CN([C@@H]3O[C@@H]4COP([O-])(=O)O[C@H]4[C@H]3O)C2=NC=N1
Appearance (Color)
White to pale cream
Form
Crystals or powder or crystalline powder
Assay from Suppliers CofA
≥98.5%

Description

Adenosine 3?,5?-cyclic monophosphate sodium salt monohydrate is used as an activator of protein kinase A (PKA) . A key regulator of many cellular reactions. Released in response to hormonal stimulation. . Directly activates Epac, a Rap1 guanine-nucleotide exchange factor. Acts as a second messenger in signal transduction mechanisms. Promotes apoptosis in resting human B lymphocytes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Adenosine 3′,5′-cyclic monophosphate sodium salt monohydrate is used as an activator of protein kinase A (PKA) . A key regulator of many cellular reactions. Released in response to hormonal stimulation. . Directly activates Epac, a Rap1 guanine-nucleotide exchange factor. Acts as a second messenger in signal transduction mechanisms. Promotes apoptosis in resting human B lymphocytes.

Solubility
Soluble in water (50 mg/ml).

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text