Danazol is a synthetic gonadotropin inhibitor which has a preventive effect on estrogen -related endometrial carcinogenesis in mice. Danazol therapy has also shown regression of established mammary carcinoma in rats. This medication is used in women to treat pelvic pain and infertility due to a certain uterus disorder (endometriosis) and also to treat breast pain/tenderness/nodules due to a certain breast condition (fibrocystic breast disease). It is also used in both men and women to prevent swelling of the abdomen/arms/legs/face/airway due to a certain congenital disease
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Partly soluble in water. Soluble in chloroform (25 mg/ml), acetone, acetonitrile, and ethanol. 17beta-Hydroxy-2,4,17a-pregnadien-20-yno[2,3-d]isoxazole and 2,4,17a-Pregnadien-20-yno[2,3-d]isoxazol-17-ol are the synonym of this compound.
Notes
Store at room temperature. Incompatible with oxidizing agents.
RUO – Research Use Only
Gary G. Liversidge.; Kenneth C. Cundy. Particle size reduction for improvement of oral bioavailability of hydrophobic drugs: I. Absolute oral bioavailability of nanocrystalline danazol in beagle dogs.Int. J. Pharm. 1995, 125 (1),91-97 .