Danazol
Danazol
Danazol
Thermo Scientific Chemicals

Danazol

CAS: 17230-88-5 | C22H27NO2 | 337.463 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
100 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J62200.ME
also known as J62200-ME
Price (USD)
182.65
Special offer
Online exclusive
Ends: 26-Sep-2025
226.00
Save 43.35 (19%)
Each
Add to cart
Quantity:
500 mg
Request bulk or custom format
Price (USD)
182.65
Special offer
Online exclusive
Ends: 26-Sep-2025
226.00
Save 43.35 (19%)
Each
Add to cart
Specifications
CAS17230-88-5
ChEBICHEBI:4315
Chemical Name or MaterialDanazol
ColorWhite to Yellow
Formula Weight337.46
View more
Danazol is a synthetic gonadotropin inhibitor which has a preventive effect on estrogen -related endometrial carcinogenesis in mice. Danazol therapy has also shown regression of established mammary carcinoma in rats. This medication is used in women to treat pelvic pain and infertility due to a certain uterus disorder (endometriosis) and also to treat breast pain/tenderness/nodules due to a certain breast condition (fibrocystic breast disease). It is also used in both men and women to prevent swelling of the abdomen/arms/legs/face/airway due to a certain congenital disease

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Partly soluble in water. Soluble in chloroform (25 mg/ml), acetone, acetonitrile, and ethanol. 17beta-Hydroxy-2,4,17a-pregnadien-20-yno[2,3-d]isoxazole and 2,4,17a-Pregnadien-20-yno[2,3-d]isoxazol-17-ol are the synonym of this compound.

Notes
Store at room temperature. Incompatible with oxidizing agents.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only
Gary G. Liversidge.; Kenneth C. Cundy. Particle size reduction for improvement of oral bioavailability of hydrophobic drugs: I. Absolute oral bioavailability of nanocrystalline danazol in beagle dogs.Int. J. Pharm. 1995, 125 (1),91-97 .

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link