Thermo Scientific Chemicals

Tunicamycin, 95%, Thermo Scientific Chemicals

Catalog number: J62217.MA
10 mg, Each
Thermo Scientific Chemicals

Tunicamycin, 95%, Thermo Scientific Chemicals

Catalog number: J62217.MA
10 mg, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J62217.MA
also known as J62217-MA
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
11089-65-9
IUPAC Name
(2E)-N-[(2R,3R,4R,5R,6R)-6-{2-[(2R,3S,4R,5R)-5-(2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]-2-hydroxyethyl}-2-{[(2R,3R,4R,5S,6R)-3-acetamido-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-4,5-dihydroxyoxan-3-yl]-5-methylhex-2-enamide
Molecular Formula
C30H46N4O16
InChI Key
ZHSGGJXRNHWHRS-PEALBESXSA-N
SMILES
CC(C)C\C=C\C(=O)N[C@@H]1[C@@H](O)[C@@H](O)[C@@H](CC(O)[C@H]2O[C@H]([C@H](O)[C@@H]2O)N2C=CC(=O)NC2=O)O[C@@H]1O[C@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1NC(C)=O
Assay from Suppliers CofA
≥94.0%
Form
Powder

Description

Tunicamycin has been widely used in the study of glycoprotein synthesis in various biological systems. During protein glycosylation, tunicamycin is noted to be an inhibitor of the transfer of saccharide moieties to dolichol during dolichol-linked glycoprotein synthesis. Dose-dependent inhibition of DNA synthesis may be related to the alteration of glycoproteins, which thereby affects the transport of thymidine into cells. Additionally, tunicamycin has been reported to prevent cell cycle progression in primary cultures of rat glial cells, as well as inhibit lipid-mediated protein glycosylation in chick or mouse fibroblasts in a dose-dependent manner.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tunicamycin has been widely used in the study of glycoprotein synthesis in various biological systems. During protein glycosylation, tunicamycin is noted to be an inhibitor of the transfer of saccharide moieties to dolichol during d
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text