9-cis-Retinoic acid, Thermo Scientific Chemicals
9-cis-Retinoic acid, Thermo Scientific Chemicals
Thermo Scientific Chemicals

9-cis-Retinoic acid, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
5 mg
Catalog number J62219.LB0
also known as J62219-#0
Price (USD)/ Each
260.00
-
Add to cart
Quantity:
5 mg
Request bulk or custom format
Price (USD)/ Each
260.00
Add to cart
9-cis-Retinoic acid, Thermo Scientific Chemicals
Catalog numberJ62219.LB0
Price (USD)/ Each
260.00
-
Add to cart
Chemical Identifiers
CAS5300-03-8
IUPAC Name(2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraenoic acid
Molecular FormulaC20H28O2
InChI KeySHGAZHPCJJPHSC-YCNIQYBTSA-N
SMILESC\C(\C=C\C1=C(C)CCCC1(C)C)=C/C=C/C(/C)=C/C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow to yellow
Mass SpecConforms to structure
FormPowder
Proton NMRConforms to structure
9-cis-Retinoic acid is a vitamin A analog that inhibits cell proliferation and induces cell differentiation. 9-cis-Retinoic acid is an inhibitor of Cox-2 and an activator of RAR α, RAR β, RAR γ and RXR.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9-cis-Retinoic acid is a vitamin A analog that inhibits cell proliferation and induces cell differentiation. 9-cis-Retinoic acid is an inhibitor of Cox-2 and an activator of RAR α, RAR β, RAR γ and RXR.

Solu
RUO – Research Use Only

General References:

  1. SA Kliewer.; K Umesono.; DJ Noonan.; RA Heyman. Convergence of 9-cis retinoic acid and peroxisome proliferator signalling pathways through heterodimer formation of their receptors. Nature. 1992, 358, 771 - 774.
  2. RA Heyman.; DJ Mangelsdorf.; JA Dyck.; RB Stein. 9-cis retinoic acid is a high affinity ligand for the retinoid X receptor. Cell. 1992, 68, (2), 397-406.