Amlodipine, 97+%, Thermo Scientific Chemicals
Amlodipine, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Amlodipine, 97+%, Thermo Scientific Chemicals

A calcium channel antagonist with potent antioxidant activity
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J62242.09
also known as J62242-09
Price (USD)/ Each
688.00
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
688.00
Add to cart
Amlodipine, 97+%, Thermo Scientific Chemicals
Catalog numberJ62242.09
Price (USD)/ Each
688.00
-
Add to cart
Specifications
Chemical Name or MaterialAmlodipine
CAS88150-42-9
Health Hazard 1H301-H318-H373
Health Hazard 2GHS H Statement
H301-H318-H373
Toxic if swallowed.
Causes serious eye damage.
May cause damage to organs through prolonged or repeated exposure.
Health Hazard 3P260-P264b-P270-P280-P301+P310-P305+P351+P338-P310-P314-P330-P501c
View more
A calcium channel antagonist with potent antioxidant activityAmlodipine is an L-type calcium channel protein inhibitor and blocker. It is used in combination with the angiotensin receptor blocker olmesartan. Further, it acts as a dihydropyridine calcium channel blocker. It is used as an anti-hypertensive. It is involved in the treatment of angina and to lower blood pressure. In addition to this, it is used to prevent chest pain and in coronary artery disease.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in dimethyl sulfoxide, ethanol, water, dimethyl formamide, acetone, chloroform, ethyl acetate and methanol.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Akamine, Y.; Uehara, H.; Miura, M.; Yasui-Furukori, N.; Uno, T. Multiple inductive effects of carbamazepine on combined therapy with paliperidone and amlodipine. J. Clin. Pharm. Ther. 2015, 40 (4), 480-482.
  2. Norouzi, P.; Gupta, V. K.; Larijani, B.; Rasoolipour, S.; Faridbod, F.; Ganjali, M. R. Coulometric differential FFT admittance voltammetry determination of Amlodipine in pharmaceutical formulation by nano-composite electrode. Talanta 2015, 131, 577-584.