Thermo Scientific Chemicals

1H-[1,2,4]Oxadiazolo[4,3-a]-quinoxalin-1-one, 98+%, Thermo Scientific Chemicals

Catalog number: J62250.MF
50 mg, Each
Thermo Scientific Chemicals

1H-[1,2,4]Oxadiazolo[4,3-a]-quinoxalin-1-one, 98+%, Thermo Scientific Chemicals

Catalog number: J62250.MF
50 mg, Each
Quantity
Catalog number: J62250.MF
also known as J62250-MF
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
41443-28-1
IUPAC Name
1H-[1,2,4]oxadiazolo[4,3-a]quinoxalin-1-one
Molecular Formula
C9H5N3O2
InChI Key
LZMHWZHOZLVYDL-UHFFFAOYSA-N
SMILES
O=C1ON=C2C=NC3=CC=CC=C3N12
Appearance (Color)
Pale yellow to yellow
Assay from Suppliers CofA
≥98.0%
Form
Powder

Description

Selective inhibitor of nitric oxide-sensitive guanylyl cyclase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text