Thermo Scientific Chemicals

Pirenzepine dihydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: J62252.03
1 g, Each
Thermo Scientific Chemicals

Pirenzepine dihydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: J62252.03
1 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J62252.03
also known as J62252-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
29868-97-1
IUPAC Name
dihydrogen 2-[2-(4-methylpiperazin-1-yl)acetyl]-2,4,9-triazatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-10-one dichloride
Molecular Formula
C19H23Cl2N5O2
InChI Key
FFNMBRCFFADNAO-UHFFFAOYSA-N
SMILES
[H+].[H+].[Cl-].[Cl-].CN1CCN(CC(=O)N2C3=CC=CC=C3C(=O)NC3=CC=CN=C23)CC1
Form
Crystals or powder or crystalline powder
Appearance (Color)
White to pale yellow
Assay from Supplier's CofA
≥98.5%

Description

A M1 muscarinic receptor antagonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text