Thermo Scientific Chemicals

(R)-(-)-2-Oxothiazolidine-4-carboxylic acid, Thermo Scientific Chemicals

Catalog number: J62254.03
1 g, Each
Thermo Scientific Chemicals

(R)-(-)-2-Oxothiazolidine-4-carboxylic acid, Thermo Scientific Chemicals

Catalog number: J62254.03
1 g, Each
Quantity
Catalog number: J62254.03
also known as J62254-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
19771-63-2
IUPAC Name
(4R)-2-oxo-1,3-thiazolidine-4-carboxylic acid
Molecular Formula
C4H5NO3S
InChI Key
BMLMGCPTLHPWPY-REOHCLBHSA-N
SMILES
OC(=O)[C@@H]1CSC(=O)N1
Appearance (Color)
White
Assay from Supplier's CofA
≥90%
Form
Crystals or crystalline powder
Proton NMR
Conforms to structure

Description

Reagent for hepatic glutathione studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text