Thermo Scientific Chemicals

5-Iodouridine, 96%, Thermo Scientific Chemicals

Catalog number: J62259.06
5 g, Each
Thermo Scientific Chemicals

5-Iodouridine, 96%, Thermo Scientific Chemicals

Catalog number: J62259.06
5 g, Each
Quantity
Catalog number: J62259.06
also known as J62259-06
Price (USD)
Quantity
-
Have Questions?

Description

5-Iodouridine has shown to enhance the effect of gamma irradiation in hamster cells. It is used as catalytic agent, petrochemical additive, used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Iodouridine has shown to enhance the effect of gamma irradiation in hamster cells. It is used as catalytic agent, petrochemical additive, used in organic synthesis.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text