Thermo Scientific Chemicals

Ethidium bromide soln., 10mg/ml, Thermo Scientific Chemicals

Catalog number: J62282.AB
10 mL, Each
Thermo Scientific Chemicals

Ethidium bromide soln., 10mg/ml, Thermo Scientific Chemicals

Catalog number: J62282.AB
10 mL, Each
Quantity
Catalog number: J62282.AB
also known as J62282-AB
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1239-45-8
IUPAC Name
3,8-diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide
Molecular Formula
C21H20BrN3
InChI Key
ZMMJGEGLRURXTF-UHFFFAOYSA-N
SMILES
[Br-].CC[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12
Appearance (Color)
Dark red
Form
Liquid
Concentration
10 mg/mL

Description

Ethidium bromide has been used in a number of fluorimetric assays for nucleic acids.1,2,3 It has been shown to bind to single-stranded DNA (although not as strongly) and triple-stranded DNA.4 Because of its ability to bind to DNA, EtBr is an inhibitor of DNA polymerase. Ethidium bromide intercalates double-stranded DNA and RNA and acts as a frameshift mutagen. It can also be used in conjunction with acridine orange to differentiate between viable, apoptotic and necrotic cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text