Thermo Scientific Chemicals

L-Cystine dihydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: J62292.30
250 g, Each
Thermo Scientific Chemicals

L-Cystine dihydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: J62292.30
250 g, Each
Quantity
Catalog number: J62292.30
also known as J62292-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
30925-07-6
IUPAC Name
dihydrogen (2R)-2-amino-3-[(2-amino-2-carboxyethyl)disulfanyl]propanoic acid dichloride
Molecular Formula
C6H14Cl2N2O4S2
InChI Key
HHGZUQPEIHGQST-VBCXBIGUSA-N
SMILES
[H+].[H+].[Cl-].[Cl-].NC(CSSC[C@H](N)C(O)=O)C(O)=O
Form
Crystalline powder or powder
Impurity content
≤0.03% (Ammonium)
Comment
From a non-animal source
Heavy Metals
≤10 ppm (as Pb)
Impurity content
≤0.03% (Sulfate)

Description

Non-animal sourced L-cystine is used as a cell culture media component for the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Non-animal sourced L-cystine is used as a cell culture media component for the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies.

Solubility
Soluble in hydrochloric acid. (0.5 M HCl: 50 mg/mL) Soluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text