Thermo Scientific Chemicals

Berberine chloride, Thermo Scientific Chemicals

Catalog number: J62311.06
5 g, Each
Thermo Scientific Chemicals

Berberine chloride, Thermo Scientific Chemicals

Catalog number: J62311.06
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J62311.06
also known as J62311-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
633-65-8
IUPAC Name
16,17-dimethoxy-5,7-dioxa-13λ⁵-azapentacyclo[11.8.0.0²,¹⁰.0⁴,⁸.0¹⁵,²⁰]henicosa-1(21),2,4(8),9,13,15,17,19-octaen-13-ylium chloride
Molecular Formula
C20H18ClNO4
InChI Key
VKJGBAJNNALVAV-UHFFFAOYSA-M
SMILES
[Cl-].COC1=CC=C2C=C3C4=CC5=C(OCO5)C=C4CC[N+]3=CC2=C1OC
Appearance (Color)
Yellow to orange
Assay from Supplier's CofA
≥88.0%
Form
Powder

Description

Berberine is an isoqinoline alkaloid shown to have a chemopreventive property against colon tumor formation by inhibiting the enzyme Cox-2 (cyclooxygenase-2) which is abundantly expressed in colon cancer cells. Also inhibits c-Jun (Activator Protein 1), a transcription factor that plays a critical role in inflammation and carcinogenesis. Berberine upregulates the expression of Pgp in hepatoma cells. Treatment with berberine potentially results in the reduced accumulation of chemotherapeutic drugs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in dimethyl sulfoxide (74 mg/ml at 25°C), ethanol (<1mg/mL at 25°C), water (<1mg/mL at 25°C), and methanol.

Notes
Store at -20°C. Incompatible with oxidizing agents. Protect from heat.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text