Thermo Scientific Chemicals

Valinomycin, 90+%, Thermo Scientific Chemicals

Catalog number: J62312.MC
100 mg, Each
Thermo Scientific Chemicals

Valinomycin, 90+%, Thermo Scientific Chemicals

Catalog number: J62312.MC
100 mg, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J62312.MC
also known as J62312-MC
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2001-95-8
IUPAC Name
(3S,6S,9R,12R,15S,18S,21R,24R,27S,30S,36R)-6,18,30-trimethyl-3,9,12,15,21,24,27,33,36-nonakis(propan-2-yl)-1,7,13,19,25,31-hexaoxa-4,10,16,22,28,34-hexaazacyclohexatriacontane-2,5,8,11,14,17,20,23,26,29,32,35-dodecone
Molecular Formula
C54H90N6O18
InChI Key
FCFNRCROJUBPLU-UYBNATROSA-N
SMILES
CC(C)[C@@H]1NC(=O)[C@H](C)OC(=O)C(NC(=O)[C@H](OC(=O)[C@@H](NC(=O)[C@H](C)OC(=O)[C@H](NC(=O)[C@H](OC(=O)[C@@H](NC(=O)[C@H](C)OC(=O)[C@H](NC(=O)[C@H](OC1=O)C(C)C)C(C)C)C(C)C)C(C)C)C(C)C)C(C)C)C(C)C)C(C)C
Appearance (Color)
White
Assay from Supplier's CofA
≥90.0%
Identification (FTIR)
Conforms
Form
Powder or crystals or crystalline powder

Description

Valinomycin, is used as a potassium ionophore shown to induce cell death. Due to the ion-selective nature of valinomycin, it is used in ion-selective electrodes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Valinomycin, is used as a potassium ionophore shown to induce cell death. Due to the ion-selective nature of valinomycin, it is used in ion-selective electrodes.

Solubility
Soluble in DMSO at 10mg/ml. Insoluble in w
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text