Thermo Scientific Chemicals

Rhodamine 6G

Catalog number: J62315.14
25 g, Each
Thermo Scientific Chemicals

Rhodamine 6G

Catalog number: J62315.14
25 g, Each
Quantity
Catalog number: J62315.14
also known as J62315-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
989-38-8
IUPAC Name
hydrogen ethyl 2-[6-(ethylamino)-3-(ethylimino)-2,7-dimethyl-3H-xanthen-9-yl]benzoate chloride
Molecular Formula
C28H31ClN2O3
InChI Key
VYXSBFYARXAAKO-UHFFFAOYSA-N
SMILES
[H+].[Cl-].CCNC1=C(C)C=C2C(OC3=CC(=NCC)C(C)=CC3=C2C2=CC=CC=C2C(=O)OCC)=C1
Form
Powder or crystals or crystalline powder
UV/Vis
UV Ratios: Absorbance at 526-528: 0.9-1.0
Appearance (Color)
Maroon to red to red-brown
Assay (unspecified)
97% - 103%

Description

Rhodamine dyes are used extensively in biotechnology applications such as fluorescence microscopy, flow cytometry,fluorescence correlation spectroscopy and ELISA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Rhodamine dyes are used extensively in biotechnology applications such as fluorescence microscopy, flow cytometry,fluorescence correlation spectroscopy and ELISA.

Solubility
Soluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents. Ensure good ventilation.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text