Thermo Scientific Chemicals

beta-Nicotinamide adenine dinucleotide, 97%, Thermo Scientific Chemicals

Catalog number: J62337.14
25 g, Each
Thermo Scientific Chemicals

beta-Nicotinamide adenine dinucleotide, 97%, Thermo Scientific Chemicals

Catalog number: J62337.14
25 g, Each
Quantity
Catalog number: J62337.14
also known as J62337-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
53-84-9
IUPAC Name
1-[(2R,3R,4S,5R)-5-{[({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl phosphonato)oxy]methyl}-3,4-dihydroxyoxolan-2-yl]-3-carbamoyl-1λ⁵-pyridin-1-ylium
Molecular Formula
C21H27N7O14P2
InChI Key
BAWFJGJZGIEFAR-WIWLTUSXNA-N
SMILES
NC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP(O)(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O
Water Content (Karl Fischer Titration)
? 8%
lambda-max (UV-Vis)
? 340nM: (6.3 ± 0.2) X103 L/mole/cm (at pH 10, reduced with alcohol dehyrogenase )
Spectral Ratio
A280/A260: 0.21 ± 0.02 (at pH 7.5)
Assay (special)
? 97%
Appearance (Color)
Pale cream to pail yellow

Description

NAD+, Free Acid is a major electron acceptor molecule in biological oxidation reactions. It is also used as a catalyst intermediate and several enzymes in a multistep system, known as enzyme cycling,.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
NAD+, Free Acid is a major electron acceptor molecule in biological oxidation reactions. It is also used as a catalyst intermediate and several enzymes in a multistep system, known as enzyme cycling,.

Solubility
Soluble in water at 50mg/ml

Notes
b-NAD is very hygroscopic and should be stored desiccated. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature -20°C.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text